Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,2,6,6-tetramethyl-4-piperidyl) Sebacate 98.0+%, TCI America™
SDP

Catalog No. S0448250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0448250G 250 g
S04485G 5 g
S044825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S0448250G Supplier TCI America Supplier No. S0448250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52829-07-9
Molecular Formula C28H52N2O4
Molecular Weight (g/mol) 480.73
MDL Number MFCD00134709
InChI Key RWMYPXKVMUFMKS-UHFFFAOYSA-N
Synonym bis 2,2,6,6-tetramethyl-4-piperidyl sebacate, bis 2,2,6,6-tetramethylpiperidin-4-yl decanedioate, tinuvin 770, sanol, sumisorb 577, tinuvin 770df, tinuvin 770ls, sanol 770, sanol ls 700, sanol ls 770
PubChem CID 164282
IUPAC Name bis(3,3,5,5-tetramethylpiperidin-4-yl) decanedioate
SMILES CC1(C)CNCC(C)(C)C1OC(=O)CCCCCCCCC(=O)OC1C(C)(C)CNCC1(C)C

Specifications

Melting Point 83°C
Color White
Quantity 250 g
UN Number 2811
Formula Weight 480.73
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,2,6,6-tetramethyl-4-piperidyl) Sebacate
EINECSNumber (5)-3732
RTECSNumber HD8315000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.