Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,4-pentanedionato)zinc(II) 96.0+%, TCI America™
SDP

Catalog No. Z0002100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0002100G 100 g
Z000225G 25 g
Z0002500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Z0002100G Supplier TCI America Supplier No. Z0002100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14024-63-6
Molecular Formula C10H14O4Zn
Molecular Weight (g/mol) 263.60
MDL Number MFCD00000035
InChI Key CYDXJXDAFPJUQE-FDGPNNRMSA-L
Synonym zinc acetylacetonate hydrate, zinc 2,4-pentanedionate, hydrate, zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
PubChem CID 131675103
IUPAC Name zinc(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Formula Weight 263.60
Percent Purity ≥96.0% (T)
Chemical Name or Material Bis(2,4-pentanedionato)zinc(II)
EINECSNumber (2)-2181
RTECSNumber ZH0917500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.