Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-aminophenyl) ether, 98%

Catalog No. AAH5539606
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 24878-25-9 | C12H12N2O | 200.241 g/mol

It is catalytic reagent and used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is catalytic reagent and used in pharmaceutical industry.

Solubility
Slightly soluble in water.

Notes
Store at low temperature, away from light and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24878-25-9
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD00792533
InChI Key GOJFAKBEASOYNM-UHFFFAOYSA-N
Synonym 2,2'-oxydianiline, 2-2-aminophenoxy aniline, 2,2 inverted exclamation marka-oxydianiline, aminophenyl ether, o,o'-oxydianiline, 2-aminophenyl ether, bis 2-aminophenyl ether, 2,2'-oxybis benzenamine, benzenamine,2,2'-oxybis, 2,2'-diaminodiphenyl ether
PubChem CID 458824
IUPAC Name 2-(2-aminophenoxy)aniline
SMILES C1=CC=C(C(=C1)N)OC2=CC=CC=C2N

Specifications

Melting Point 61°C to 64°C
Color Brown to White
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 200.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bis(2-aminophenyl) ether
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.