Alfa Aesar™ Bis(2-aminophenyl) ether, 98%

Manufacturer: Alfa Aesar™ H55396MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55396MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24878-25-9
    Formula Weight 200.24
    MDL Number MFCD00792533
    Synonym 2-Aminophenyl ether; 2,2'-Oxydianiline
    Chemical Name or Material Bis(2-aminophenyl) ether

    Specifications

    Formula Weight 200.24
    Melting Point 61° to 64°C
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    MDL Number MFCD00792533
    Color White to pale brown
    Synonym 2-Aminophenyl ether; 2,2'-Oxydianiline
    Health Hazard 1 Warning
    CAS 24878-25-9
    Quantity 250mg
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Bis(2-aminophenyl) ether
    Assay 98%

    Description

    Applications
    It is catalytic reagent and used in pharmaceutical industry.

    Solubility
    Slightly soluble in water.

    Notes
    Store at low temperature, away from light and moisture.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit