Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-chloroethyl)amine hydrochloride, 98%

Catalog No. ac106321000
Click to view available options
:
100g; Plastic bottle

CAS: 821-48-7 | C4H9Cl2N·HCl | 178.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 821-48-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 210°C to 215°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C4H9Cl2N·HCl
Linear Formula (ClCH2CH2)2NH·HCl
Quantity 100 g
MDL Number MFCD00012515
Synonym bis 2-chloroethyl amine hydrochloride, 2,2'-dichlorodiethylamine hydrochloride, 2-chloro-n-2-chloroethyl ethanamine hydrochloride, nor-lost hydrochlorid, nor-hn2, ethanamine, 2-chloro-n-2-chloroethyl-, hydrochloride, n,n-bis 2-chloroethyl amine hydrochloride, di-2-chloroethyl amine hydrochloride, 2,2-dichlorodiethylamine hydrochloride, bis-2-chloroethyl amine hydrochloride
Solubility Information Solubility in water: soluble in water. Other solubilities: sparingly soluble in alcohols,insoluble in toluene and acetone
InChI Key YMDZDFSUDFLGMX-UHFFFAOYSA-N
SMILES C(CCl)NCCCl.Cl
IUPAC Name 2-chloro-N-(2-chloroethyl)ethanamine;hydrochloride
Molecular Weight (g/mol) 178.49
PubChem CID 522769
Formula Weight 178.49
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-chloroethyl)amine hydrochloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-479-5
RTECSNumber IA1225000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.