Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bis(2-ethylhexyl) phthalate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(2-ethylhexyl) phthalate is used as dielectric fluid in capacitors, erasable inks and vacuum pump oils. It is also used as plasticizer. Further, it is used as a hydraulic fluid and as a solvent in glow sticks. In addition to this, it is used to give softness and flexibility to polyvinyl chloride products. It finds application in products like tablecloths, medical tubing, floor tiles, toys, shower curtains, rainwear, dolls, shoes, garden hoses, furniture upholstery, and swimming pool liners.
Solubility
Miscible with oil. Immiscible with water.
Notes
Store in a cool place. Incompatible with nitrates and strong oxidizing agents.
Bis(2-ethylhexyl) phthalate is used as dielectric fluid in capacitors, erasable inks and vacuum pump oils. It is also used as plasticizer. Further, it is used as a hydraulic fluid and as a solvent in glow sticks. In addition to this, it is used to give softness and flexibility to polyvinyl chloride products. It finds application in products like tablecloths, medical tubing, floor tiles, toys, shower curtains, rainwear, dolls, shoes, garden hoses, furniture upholstery, and swimming pool liners.
Solubility
Miscible with oil. Immiscible with water.
Notes
Store in a cool place. Incompatible with nitrates and strong oxidizing agents.

Specifications
Specifications
Melting Point | -55°C |
Density | 0.985 |
Boiling Point | 230°C (5 mmHg) |
Flash Point | 207°C (404°F) |
Odor | Odorless |
Refractive Index | 1.487 |
Quantity | 2500 g |
Beilstein | 1890696 |
Merck Index | 14,2864 |
Solubility Information | Miscible with oil. Immiscible with water. |
Show More |
Safety and Handling
Hazard Category | H350-H360FD |
Hazard Statement | GHS H Statement H360 May damage fertility or the unborn child. |
Precautionary Statement | P201-P202-P281-P308+P313-P501c |
EINECSNumber | 204-211-0 |
RTECSNumber | TI0350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction