Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) phthalate, 98+%, Thermo Scientific Chemicals

Catalog No. aaa104150c
Click to view available options
Quantity:
10,000 g
2500 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-ethylhexyl) phthalate is used as dielectric fluid in capacitors, erasable inks and vacuum pump oils. It is also used as plasticizer. Further, it is used as a hydraulic fluid and as a solvent in glow sticks. In addition to this, it is used to give softness and flexibility to polyvinyl chloride products. It finds application in products like tablecloths, medical tubing, floor tiles, toys, shower curtains, rainwear, dolls, shoes, garden hoses, furniture upholstery, and swimming pool liners.

Solubility
Miscible with oil. Immiscible with water.

Notes
Store in a cool place. Incompatible with nitrates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-81-7
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.564
MDL Number MFCD00009493
InChI Key BJQHLKABXJIVAM-UHFFFAOYSA-N
Synonym dehp, bis 2-ethylhexyl phthalate, di 2-ethylhexyl phthalate, diethylhexyl phthalate, di-sec-octyl phthalate, octyl phthalate, fleximel, octoil
PubChem CID 8343
ChEBI CHEBI:17747
IUPAC Name bis(2-ethylhexyl) benzene-1,2-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC

Specifications

Melting Point -55°C
Density 0.985
Boiling Point 230°C (5 mmHg)
Flash Point 207°C (404°F)
Odor Odorless
Refractive Index 1.487
Quantity 10,000 g
Beilstein 1890696
Merck Index 14,2864
Solubility Information Miscible with oil. Immiscible with water.
Formula Weight 390.56
Percent Purity ≥98%
Chemical Name or Material Bis (2-ethylhexyl) phthalate
Show More Show Less
Hazard Category H350-H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 204-211-0
RTECSNumber TI0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.