Alfa Aesar™ Bis(2-methoxyethyl) phthalate, 96%

Manufacturer: Alfa Aesar™ 4408918

Quantity 50g

 View more versions of this product

Catalog No. AA4408918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 117-82-8
    Molecular Formula C14H18O6
    Formula Weight 282.29
    MDL Number MFCD00042842
    Beilstein 2056929
    Chemical Name or Material Bis (2-methoxyethyl) phthalate

    Specifications

    Formula Weight 282.29
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Liquid
    Beilstein 2056929
    Flash Point 185°C (365°F)
    Health Hazard 1 Danger
    CAS 117-82-8
    Health Hazard 2 GHS H Statement
    H360
    May damage fertility or the unborn child.
    Density 1.17g/mL
    RTECS Number TI1400000
    Molecular Formula C14H18O6
    MDL Number MFCD00042842
    Color Colorless
    Boiling Point 340°C
    Refractive Index 1.503
    Quantity 50g
    Chemical Name or Material Bis (2-methoxyethyl) phthalate
    Assay 96%

    GHS H Statement
    H360
    May damage fertility or the unborn child.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit