Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-pyridylmethyl)amine 98.0+%, TCI America™
SDP

Catalog No. D222825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D222825G 25 g
D22285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D222825G Supplier TCI America Supplier No. D222825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1539-42-0
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.257
MDL Number MFCD00129044
InChI Key KXZQYLBVMZGIKC-UHFFFAOYSA-N
Synonym 2,2′C-Dipicolylamine, alpha,alpha′C-Iminodi(2-picoline)
PubChem CID 73759
IUPAC Name 1-pyridin-2-yl-N-(pyridin-2-ylmethyl)methanamine
SMILES C1=CC=NC(=C1)CNCC2=CC=CC=N2

Specifications

Color Yellow
Boiling Point 144°C
Quantity 25 g
Formula Weight 199.26
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Bis(2-pyridylmethyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.