Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-pyridylmethyl)amine 98.0+%, TCI America™
SDP

Catalog No. p-7127177
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1539-42-0
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.257
MDL Number MFCD00129044
InChI Key KXZQYLBVMZGIKC-UHFFFAOYSA-N
Synonym 2,2′C-Dipicolylamine, alpha,alpha′C-Iminodi(2-picoline)
PubChem CID 73759
IUPAC Name 1-pyridin-2-yl-N-(pyridin-2-ylmethyl)methanamine
SMILES C1=CC=NC(=C1)CNCC2=CC=CC=N2

Specifications

Color Yellow
Boiling Point 144°C
Quantity 5 g
Formula Weight 199.26
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Bis(2-pyridylmethyl)amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.