Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(3-pyridylmethyl)amine 97.0+%, TCI America™
SDP

Catalog No. D119925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D119925ML 25 mL
D11995ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D119925ML Supplier TCI America Supplier No. D119925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1656-94-6
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.26
MDL Number MFCD00038043
InChI Key FEBQXMFOLRVSGC-UHFFFAOYSA-N
PubChem CID 74257
IUPAC Name bis[(pyridin-3-yl)methyl]amine
SMILES C(NCC1=CC=CN=C1)C1=CC=CN=C1

Specifications

Color Yellow
Quantity 25 mL
Synonym 3,3′-Dipicolylamine, alpha,alpha′-Iminodi(3-picoline)
Formula Weight 199.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(3-pyridylmethyl)amine
RTECSNumber UT4397000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.