Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-aminophenyl) ether, 98%

Catalog No. p-7023358
Click to view available options
Quantity:
50 g
100 g
500 g

CAS: 101-80-4 | C12H12N2O | 200.241 g/mol

Bis(4-aminophenyl) ether is a monomer used as a cross-linking agent. It is also used in the preparation of poly(imideurea)s, polyimide silica composite membrane, which finds application in gas separation techniques. It is utilized in products such as wire enamels, coatings, adhesives, insulating varnishes, coated fabrics, flame-retardant fibers, oil sealants and retainers. Further, it is used in the preparation of polymer films for electrolyte membranes. In addition to this, it is used as an insulation for cables and printed circuits, and laminates and composite for aerospace vehicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-aminophenyl) ether is a monomer used as a cross-linking agent. It is also used in the preparation of poly(imideurea)s, polyimide silica composite membrane, which finds application in gas separation techniques. It is utilized in products such as wire enamels, coatings, adhesives, insulating varnishes, coated fabrics, flame-retardant fibers, oil sealants and retainers. Further, it is used in the preparation of polymer films for electrolyte membranes. In addition to this, it is used as an insulation for cables and printed circuits, and laminates and composite for aerospace vehicles.

Solubility
Soluble in acetone. Insoluble in water, benzene, carbon tetrachloride, ethanol and hot methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-80-4
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD00007863
InChI Key HLBLWEWZXPIGSM-UHFFFAOYSA-N
Synonym 4,4'-oxydianiline, 4,4'-diaminodiphenyl ether, 4,4-oxydianiline, 4-aminophenyl ether, 4,4'-oxybisbenzenamine, oxydianiline, 4-4-aminophenoxy aniline, diaminodiphenyl ether, benzenamine, 4,4'-oxybis, p,p'-oxydianiline
PubChem CID 7579
ChEBI CHEBI:34384
IUPAC Name 4-(4-aminophenoxy)aniline
SMILES C1=CC(=CC=C1N)OC2=CC=C(C=C2)N

Specifications

Melting Point 189°C to 192°C
Boiling Point 190°C (0.1 mmHg)
Flash Point 218°C (424°F)
Quantity 50 g
UN Number UN3077
Beilstein 475735
Solubility Information Soluble in acetone. Insoluble in water,benzene,carbon tetrachloride,ethanol and hot methanol.
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material Bis(4-aminophenyl) ether
Hazard Category H301+H311+H331-H340-H350-H360
Hazard Statement GHS H Statement
H301-H311-H331-H340-H350-H361
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 202-977-0
RTECSNumber BY7900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.