Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-bromophenyl)diphenylsilane 98.0+%, TCI America™
SDP

Catalog No. B50871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50871G 1 g
B5087200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50871G Supplier TCI America Supplier No. B50871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18733-91-0
Molecular Formula C24H18Br2Si
Molecular Weight (g/mol) 494.301
InChI Key UTEBJTKMYMQRIF-UHFFFAOYSA-N
PubChem CID 297512
IUPAC Name bis(4-bromophenyl)-diphenylsilane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br

Specifications

Melting Point 169°C
Color White
Quantity 1 g
Formula Weight 494.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-bromophenyl)diphenylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.