Learn More
Bis(4-bromophenyl) ether, 99%
Description
Bis-(4-bromophenyl)-ether has been used in the preparation of 4-[4-(diethoxyphosphoryl)phenoxy]phenyl-phosphonic acid diethyl ester. It is used for medicine and as organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 56°C to 60°C |
Color | White |
Boiling Point | 338°C to 340°C |
Quantity | 25 g |
Solubility Information | Soluble in water. |
Formula Weight | 328.01 |
Percent Purity | 99% |
Physical Form | Crystalline |
Chemical Name or Material | Bis(4-bromophenyl) ether |
Safety and Handling
EINECSNumber | 218-090-7 |
RTECSNumber | KN0175000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.