Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-diethylaminophenyl)methanol 98.0+%, TCI America™
SDP

Catalog No. B17251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17251G 1 g
B17255G 5 g
B172510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B17251G Supplier TCI America Supplier No. B17251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 134-91-8
Molecular Formula C21H30N2O
Molecular Weight (g/mol) 326.484
MDL Number MFCD00035979
InChI Key WCIQBUVXZZYFJP-UHFFFAOYSA-N
PubChem CID 67257
IUPAC Name bis[4-(diethylamino)phenyl]methanol
SMILES CCN(CC)C1=CC=C(C=C1)C(C2=CC=C(C=C2)N(CC)CC)O

Specifications

Melting Point 82°C
Color White
Quantity 1 g
Synonym 4,4′-Bis(diethylamino)benzhydrol
Formula Weight 326.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-diethylaminophenyl)methanol
EINECSNumber (4)-0883
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.