Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-hydroxyphenyl) Disulfide 98.0+%, TCI America™
SDP

Catalog No. B38271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38271G 1 g
B38275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38271G Supplier TCI America Supplier No. B38271G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15015-57-3
Molecular Formula C12H10O2S2
Molecular Weight (g/mol) 250.33
MDL Number MFCD00020164
InChI Key XGKGITBBMXTKTE-UHFFFAOYSA-N
PubChem CID 280665
IUPAC Name 4-[(4-hydroxyphenyl)disulfanyl]phenol
SMILES C1=CC(=CC=C1O)SSC2=CC=C(C=C2)O

Specifications

Melting Point 152°C
Color Yellow
Quantity 1 g
Synonym 4,4′C-Dihydroxydiphenyl Disulfide, 4,4′C-Dithiodiphenol
Formula Weight 250.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-hydroxyphenyl) Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.