Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(acetonitrile)palladium(II) chloride, 99%

Catalog No. p-68033
Change view
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC209455000 500 mg Glass bottle
AC209450050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC209455000 Supplier Thermo Scientific Chemicals Supplier No. 209455000
Only null left
Add to Cart

CAS: 14592-56-4 | C4H6Cl2N2Pd | 259.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14592-56-4
Molecular Formula C4H6Cl2N2Pd
Molecular Weight (g/mol) 259.42
MDL Number MFCD00013122
InChI Key XGPJSRXOBCGBMN-UHFFFAOYSA-L
IUPAC Name palladium(2+) bis(acetonitrile) dichloride
SMILES CC#N.CC#N.Cl[Pd++]Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 300°C
Color Brown-Rust to Orange
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3CN)2PdCl2
Quantity 500 mg
Fieser 13,33; 15,28; 16,25; 17,30; 01,1015; 03,254; 04,435
Solubility Information Solubility in water: insoluble
Formula Weight 259.4
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Bis(acetonitrile)palladium(II) chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-637-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.