Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)cobalt

Catalog No. p-7049641
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1685703 1 g
AA1685706 5 g
AA1685714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1685703 Supplier Thermo Scientific Chemicals Supplier No. 01685703
Only null left
Add to Cart

CAS: 1277-43-6 | C10H10Co | 189.123 g/mol

Bis(cyclopentadienyl)cobalt is used to catalyze the preparation of pyridines from alkynes and nitriles. It acts as a polymerization inhibitor of olefins. It is a common one-electron reducing agent in the laboratory. It reacts with carbon monoxide to prepare cobalt(I) derivative, cyclopentadienylcobalt dicarbonyl. It also serves as an internal standard in cyclic voltametry. Further, it finds use in Diels-Alder reactions. In addition to this, it is used as a paint drier and as an oxygen stripping agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1277-43-6
Molecular Formula C10H10Co
Molecular Weight (g/mol) 189.123
MDL Number MFCD00013749
InChI Key SNBRJOIYNQIWSZ-UHFFFAOYSA-N
Synonym Cobaltocene
PubChem CID 24942376
IUPAC Name cobalt(2+);cyclopenta-1,3-diene
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2]

Specifications

Melting Point 176°C to 180°C (decomposition)
Quantity 1 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 189.12
Physical Form Powder
Chemical Name or Material Bis(cyclopentadienyl)cobalt
Hazard Category H228
Hazard Statement GHS H Statement
H228-H341-H317
Flammable solid.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECSNumber 215-061-0
RTECSNumber GG0350000
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.