Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bis(cyclopentadienyl)titanium dichloride, Thermo Scientific™
Click to view available options
Quantity:
5 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under argon
Specifications
Specifications
| Melting Point | ∼285°C (decomposition) |
| Color | Red |
| Density | 1.60 g/mL |
| Odor | Odorless |
| Quantity | 5 g |
| Sensitivity | Air and moisture sensitive |
| Merck Index | 14,9482 |
| Solubility Information | It is insoluble in water |
| Formula Weight | 249 |
| Physical Form | Crystalline |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H341-H302-H332-H315-H319-H335 Suspected of causing genetic defects. Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 215-035-9 |
| RTECSNumber | XR2050000 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures; Store under Argon |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction