Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)titanium dichloride, 97%

Catalog No. AAA1145609
Click to view available options
Quantity:
10 g
50 g
250 g

Catalog No. AAA1145609

Supplier: Thermo Scientific Chemicals A1145609

Only null left
Add to Cart

CAS: 1271-19-8 | C10H10Cl2Ti-2 | 248.957 g/mol

One-pot synthesis of α-methylene-γ-butyrolactones from benzaldehydes and bromomethylacrylates. Bis-(cyclopentadienyl)-titanium dichloride alkylaluminum complexls acts as catalysts for the polymerization of ethylene. Polyethylene prepared with these catalysts. Complexes of aluminum chloride and methylaluminum dichloride with bis-(cyclopentadienyl)-titanium dichloride as catalysts for the polymerization of ethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
One-pot synthesis of α-methylene-γ-butyrolactones from benzaldehydes and bromomethylacrylates. Bis-(cyclopentadienyl)-titanium dichloride alkylaluminum complexls acts as catalysts for the polymerization of ethylene. Polyethylene prepared with these catalysts. Complexes of aluminum chloride and methylaluminum dichloride with bis-(cyclopentadienyl)-titanium dichloride as catalysts for the polymerization of ethylene.

Solubility
Slightly soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-19-8
Molecular Formula C10H10Cl2Ti-2
Molecular Weight (g/mol) 248.957
MDL Number MFCD00003723
InChI Key MKNXBRLZBFVUPV-UHFFFAOYSA-L
Synonym Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
PubChem CID 124040768
IUPAC Name cyclopenta-1,3-diene;titanium(2+);dichloride
SMILES [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]

Specifications

Melting Point ∼285°C (decomposition)
Density 1.6
Odor Odorless
Quantity 10 g
Sensitivity Air and moisture sensitive
Merck Index 14,9482
Solubility Information Slightly soluble in water
Formula Weight 249
Percent Purity 97%
Chemical Name or Material Bis (cyclopentadienyl)titanium dichloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H341-H302-H332-H315-H319-H335
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-035-9
RTECSNumber XR2050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.