Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(dicarbonylcyclopentadienyliron), 99%

Catalog No. AA3111406
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AA3111406

Supplier: Thermo Scientific Chemicals 03111406

Only null left
Add to Cart

CAS: 12154-95-9

Cyclopentadienyliron dicarbonyl dimer is a workhorse in organometallic chemistry because it is inexpensive and rugged. It is also used as a pharmaceutical intermediates and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12154-95-9
MDL Number MFCD00135625
Synonym Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyl-di-iron tetracarbonyl

Specifications

Melting Point 194°C (decomposition)
Color Red
Quantity 5 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Solubility Information It is readily soluble in moderate polar organic solvents such as chloroform and pyridine,but less soluble in carbon tetrachloride and carbon disulfide. It is sparingly soluble in water.
Formula Weight 353.93
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Bis(dicarbonylcyclopentadienyliron)
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H228-H300-H310-H330
Flammable solid.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 235-276-3
TSCA No
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.