Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(diethylamino)chlorophosphine, 94%

Catalog No. AAB2101914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2101914 25 g
AAB2101906 5 g
AAB2101922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2101914 Supplier Thermo Scientific Chemicals Supplier No. B2101914
Add to Cart

CAS: 685-83-6 | C8H20ClN2P | 210.686 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 685-83-6
Molecular Formula C8H20ClN2P
Molecular Weight (g/mol) 210.686
MDL Number MFCD00015493
InChI Key JVEHJSIFWIIFHM-UHFFFAOYSA-N
Synonym bis diethylamino chlorophosphine, bis diethylamino phosphorchloridite, n,n,n',n'-tetraethylphosphorodiamidous chloride, bdapcd, chloro diethylamino phosphanyl diethylamine, pubchem6475, phosphorodiamidous chloride, tetraethyl, chlorobis diethylamino phosphine, bisdiethylamino-chloro-phosphine, chloro-bis diethylamino-phosphine
PubChem CID 2733351
IUPAC Name N-[chloro(diethylamino)phosphanyl]-N-ethylethanamine
SMILES CCN(CC)P(N(CC)CC)Cl

Specifications

Density 1.002
Boiling Point 55°C to 57°C (0.2 mmHg)
Flash Point 101°C (214°F)
Odor Acrid
Linear Formula [(C2H5)2N]2PCl
Quantity 25 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Formula Weight 210.69
Percent Purity 94%
Chemical Name or Material Bis(diethylamino)chlorophosphine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.