Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(diethylamino)dimethylsilane 98.0+%, TCI America™
SDP

Catalog No. B549625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B549625ML 25 mL
B54965ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B549625ML Supplier TCI America Supplier No. B549625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4669-59-4
Molecular Formula C10H26N2Si
Molecular Weight (g/mol) 202.417
MDL Number MFCD00039879
InChI Key XIFOKLGEKUNZTI-UHFFFAOYSA-N
Synonym N,N,N′,N′-Tetraethyl-1,1-dimethylsilanediamine
PubChem CID 78399
IUPAC Name N-[diethylamino(dimethyl)silyl]-N-ethylethanamine
SMILES CCN(CC)[Si](C)(C)N(CC)CC

Specifications

Color Yellow
Boiling Point 193°C
Quantity 25 mL
UN Number 1993
Formula Weight 202.42
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(diethylamino)dimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.