Alfa Aesar™ Bis(dimethylamino)chlorophosphine

Manufacturer: Alfa Aesar™ 3030703

Quantity 1g

 View more versions of this product

Catalog No. AA3030703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3348-44-5
    Linear Formula [(CH3)2N]2PCl
    Formula Weight 154.58
    MDL Number MFCD00014862
    UN Number UN3265
    Chemical Name or Material Bis(dimethylamino)chlorophosphine

    Specifications

    Formula Weight 154.58
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310-P402
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Physical Form Liquid
    Refractive Index 1.5010
    Color Colorless
    Quantity 1g
    Odor Pungent
    Storage Note 1
    • Packaged under argon
    Linear Formula [(CH3)2N]2PCl
    MDL Number MFCD00014862
    CAS 3348-44-5
    UN Number UN3265
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Health Hazard 1 Danger
    Sensitivity Air & Moisture Sensitive
    Chemical Name or Material Bis(dimethylamino)chlorophosphine

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310-P402
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit