Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(hexafluoroacetylacetonato)magnesium(II) Hydrate, TCI America™
SDP

Catalog No. H05561G
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 19648-85-2
Molecular Formula C10H6F12MgO6
Molecular Weight (g/mol) 474.438
MDL Number MFCD00063975
InChI Key CDOUVTSJWSMSKS-LJDKTGGESA-L
Synonym Magnesium(II) Hexafluoroacetylacetonate, Hexafluoroacetylacetono Magnesium(II) Salt
PubChem CID 91927756
IUPAC Name magnesium;(Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate;dihydrate
SMILES C(=C(C(F)(F)F)[O-])C(=O)C(F)(F)F.C(=C(C(F)(F)F)[O-])C(=O)C(F)(F)F.O.O.[Mg+2]

Specifications

Quantity 1 g
Formula Weight 438.41
Physical Form Crystalline Powder
Chemical Name or Material Bis(hexafluoroacetylacetonato)magnesium(II) Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.