Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(methylcyclopentadienyl)niobium dichloride, 97%, Thermo Scientific Chemicals

Catalog No. AA4439306
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61374-51-4
Molecular Formula C12H14Cl2Nb
MDL Number MFCD03427138
Synonym bis methylcyclopentadienyl niobium dichloride, acmc-1b8dp, bis 2-methylcyclopenta-1,3-dien-1-yl niobiumbis ylium dichloride, niobium,dichlorobis 1,2,3,4,5-h-1-methyl-2,4-cyclopentadien-1-yl

Specifications

Melting Point 180°C (decomposition)
Color Brown
Quantity 5 g
UN Number UN3261
Sensitivity Air & Moisture Sensitive
Formula Weight 322.05
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Bis (methylcyclopentadienyl)niobium dichloride
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.