Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(methyldiphenylphosphine)palladium(II) Dichloride 96.0+%, TCI America™
SDP

Catalog No. B21615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21615G 5 g
B21611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21615G Supplier TCI America Supplier No. B21615G

Chemical Identifiers

CAS 52611-08-2
Molecular Formula C26H26Cl2P2Pd
Molecular Weight (g/mol) 577.762
MDL Number MFCD00216594
InChI Key GTSFBKGHQLDFHU-UHFFFAOYSA-L
Synonym Dichlorobis(methyldiphenylphosphine)palladium(II)
PubChem CID 6102084
IUPAC Name dichloropalladium;methyl(diphenyl)phosphane
SMILES CP(C1=CC=CC=C1)C2=CC=CC=C2.CP(C1=CC=CC=C1)C2=CC=CC=C2.Cl[Pd]Cl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 577.76
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(methyldiphenylphosphine)palladium(II) Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.