Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(methylenedithio)tetrathiafulvalene [Organic Electronic Material], TCI America™
SDP

Catalog No. B1218100MG
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 68550-20-9
Molecular Formula C8H4S8
Molecular Weight (g/mol) 356.6
MDL Number MFCD00059697
InChI Key YAHVGMRXXZDSOK-UHFFFAOYSA-N
Synonym BMDT-TTF
PubChem CID 13740883
IUPAC Name 5-([1,3]dithiolo[4,5-d][1,3]dithiol-5-ylidene)-[1,3]dithiolo[4,5-d][1,3]dithiole
SMILES C1SC2=C(S1)SC(=C3SC4=C(S3)SCS4)S2

Specifications

Color Brown-Red
Quantity 100 mg
Formula Weight 356.60
Physical Form Crystalline Powder
Chemical Name or Material Bis(methylenedithio)tetrathiafulvalene [Organic Electronic Material]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.