Learn More
Bis(tri-o-tolylphosphine)palladium(0), Pd 14.9%
Description
Bis(tri-o-tolylphosphine)palladium(0) is used as a catalyst for palladium-catalyzed cross-coupling reactions, Kumada coupling and Heck coupling. It is also used in the amination and arylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 220°C to 225°C |
Color | White to Yellow |
Assay Percent Range | Pd 14.9% |
Quantity | 1 g |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 715.16 |
Physical Form | Crystalline |
Chemical Name or Material | Bis(tri-o-tolylphosphine)palladium(0) |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.