Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(trimethylsilyl)sulfide, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3385-94-2
C6H18SSi2
178.45
MFCD00014851
RLECCBFNWDXKPK-UHFFFAOYSA-N
bis trimethylsilyl sulfide,hexamethyldisilthiane,hexamethyldisilathiane,disilathiane, hexamethyl,1,1,1,3,3,3-hexamethyldisilathiane,disilthiane, hexamethyl,disilathiane, 1,1,1,3,3,3-hexamethyl,thiobis trimethylsilane,acmc-1ckj8
76920
trimethyl(trimethylsilylsulfanyl)silane
C[Si](C)(C)S[Si](C)(C)C
3385-94-2
C6H18SSi2
178.45
MFCD00014851
RLECCBFNWDXKPK-UHFFFAOYSA-N
bis trimethylsilyl sulfide,hexamethyldisilthiane,hexamethyldisilathiane,disilathiane, hexamethyl,1,1,1,3,3,3-hexamethyldisilathiane,disilthiane, hexamethyl,disilathiane, 1,1,1,3,3,3-hexamethyl,thiobis trimethylsilane,acmc-1ckj8
76920
trimethyl(trimethylsilylsulfanyl)silane
C[Si](C)(C)S[Si](C)(C)C
Specific Gravity 0.95
Quantity 5g
Infrared Spectrum Authentic
Refractive Index 1.4555 to 1.4585
Flash Point 26°C
Boiling Point 163.0°C (170.0 mmHg)
Density 0.9500g/mL
Color Colorless to Yellow
Packaging Glass bottle
Assay Percent Range 94% min. (GC)
Linear Formula S(Si(CH3)3)2
Solubility Information Solubility in water: .. Other solubilities: soluble in toluene,ethers,hydrocarbons
Formula Weight 178.45
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Bis(trimethylsilyl)sulfide, 95%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 222-201-4