Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(triphenylphosphine)palladium(II) chloride, for analysis, 15% Pd, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13965-03-2
C36H30Cl2P2Pd
701.89
TUPQTEGWNDAOKM-UHFFFAOYSA-L
bis triphenylphosphine palladium ii dichloride,bis triphenylphosphine palladium ii chloride,dichlorobis triphenylphosphine palladium,bis triphenylphosphine palladium chloride,bis triphenylphosphine palladiuml ii dichloride,palladium 2+ ; triphenylphosphane; dichloride,trans-dichlorobis triphenylphosphine palladium ii 1g
131664180
ethane;methane;palladium(2+);triphenylphosphane;dichloride
C.CC.CC.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Cl-].[Pd+2]
13965-03-2
C36H30Cl2P2Pd
701.89
TUPQTEGWNDAOKM-UHFFFAOYSA-L
bis triphenylphosphine palladium ii dichloride,bis triphenylphosphine palladium ii chloride,dichlorobis triphenylphosphine palladium,bis triphenylphosphine palladium chloride,bis triphenylphosphine palladiuml ii dichloride,palladium 2+ ; triphenylphosphane; dichloride,trans-dichlorobis triphenylphosphine palladium ii 1g
131664180
ethane;methane;palladium(2+);triphenylphosphane;dichloride
C.CC.CC.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Cl-].[Pd+2]
Melting Point 260°C
Infrared Spectrum Authentic
Quantity 25g
Color Yellow
Packaging Glass bottle
Linear Formula [(C6H5)3P]2PdCl2
Fieser 06,59; 07,95; 08,151; 17,111
Solubility Information Solubility in water: insoluble.
Formula Weight 701.89
Physical Form Powder
Percent Purity 15% Pd
Grade Analysis
Chemical Name or Material Bis(triphenylphosphine)palladium(II) chloride
Show More Show Less

GHS H Statement
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

Warning

EINECSNumber : 237-744-2