Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(triphenyltin) oxide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass Bottle
1262-21-1
C36H30OSn2
716.055
MFCD00003005
MUHFQLVFMFDJOK-UHFFFAOYSA-N
bis triphenyltin oxide,distannoxane, hexaphenyl,fentin-oxide,fentin oxide,bis triphenylzinn-oxid,bis triphenyl tin oxide,muhfqlvfmfdjok-uhfffaoysa,triphenyl triphenylstannyloxy stannane,triphenyl triphenylstannyl oxy stannane
16683296
triphenyl(triphenylstannyloxy)stannane
C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)(C3=CC=CC=C3)O[Sn](C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6
1262-21-1
C36H30OSn2
716.055
MFCD00003005
MUHFQLVFMFDJOK-UHFFFAOYSA-N
bis triphenyltin oxide,distannoxane, hexaphenyl,fentin-oxide,fentin oxide,bis triphenylzinn-oxid,bis triphenyl tin oxide,muhfqlvfmfdjok-uhfffaoysa,triphenyl triphenylstannyloxy stannane,triphenyl triphenylstannyl oxy stannane
16683296
triphenyl(triphenylstannyloxy)stannane
C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)(C3=CC=CC=C3)O[Sn](C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6
Color White
Packaging Glass Bottle
Quantity 25g
Infrared Spectrum Authentic
Melting Point 119.0°C to 123.0°C
Assay Percent Range 98%
Linear Formula [(C6H5)3Sn]2O
Formula Weight 716.02
Physical Form Crystalline Powder and/or Crystals
Percent Purity 98%
Chemical Name or Material Bis(triphenyltin) oxide
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 215-025-4