Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(vinylsulfonyl)methane 96.0+%, TCI America™
SDP

Catalog No. B255025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B255025G 25 g
B25505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B255025G Supplier TCI America Supplier No. B255025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3278-22-6
Molecular Formula C5H8O4S2
Molecular Weight (g/mol) 196.24
MDL Number MFCD03265421
InChI Key IJHIIHORMWQZRQ-UHFFFAOYSA-N
PubChem CID 76764
IUPAC Name (ethenesulfonyl)methanesulfonylethene
SMILES C=CS(=O)(=O)CS(=O)(=O)C=C

Specifications

Melting Point 62°C
Color White
Quantity 25 g
Formula Weight 196.24
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(vinylsulfonyl)methane
EINECSNumber (2)-3734
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.