Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bisabolene (so called) [for Fragrance], TCI America™
SDP

Catalog No. B141325ML
Click to view available options
Quantity:
25 mL

Chemical Identifiers

Molecular Formula C15H24
MDL Number MFCD00129080

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 204.36
Physical Form Liquid
Chemical Name or Material Bisabolene (so called) [for Fragrance]
RTECSNumber GW6060000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.