Learn More
Stoelting™ Bishop-Harmon Forcep
SDP
Micro tweezers are useful in a variety of different applications. Common uses include micro dissection of tissue and vessels, and processing tissue in electron microscopy procedures.
$429.50 - $522.00
Specifications
Model Fine
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
10-001-199
SDP
View Documents
Stoelting™
5210275
3.25 in., Cross Serrated; 0.5 mm
Each for $429.50
Only null left
Add to Cart
10-001-200
SDP
View Documents
Stoelting™
5210276
3.25 in., 1 x 2.3
Each for $522.00
Only null left
Add to Cart
Description
Description
Specifications
Specifications
Fine
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.