Learn More
Bismuth 2-ethylhexanoate
Description
Bismuth 2-ethylhexanoate is used as an industrial lubricant and dessicant. It is also used for synthesizing of urethane foam, urethane coating and other urethane products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 25 g |
| Solubility Information | Not miscible with water. |
| Formula Weight | 638.61 |
| Physical Form | Viscous Liquid |
| Chemical Name or Material | Bismuth 2-ethylhexanoate |
Safety and Handling
| Hazard Category | H361d |
| Hazard Statement | GHS H Statement H361 Suspected of damaging fertility or the unborn child. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 267-499-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.