Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth carbonate oxide, 98.5% min

Catalog No. AA3321636
Click to view available options
Quantity:
100 g
500 g

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes | CAS: 5892-10-4 | CBi2O5 | 509.97 g/mol

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazesBismuth carbonate oxide is used in ceramic glazes and pearly coatings on plastics. It is also used as an antacid and anti-diarrheal in medicinal chemistry. Further, it is used as a filler in radiopaque catheters. It is also employed in fireworks and to make Dragon's eggs. In addition to this, it is made into nanotube arrays that exhibit antibacterial properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5892-10-4
Molecular Formula CBi2O5
Molecular Weight (g/mol) 509.97
MDL Number MFCD00010877
InChI Key MGLUJXPJRXTKJM-UHFFFAOYSA-L
Synonym bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
PubChem CID 16683095
ChEBI CHEBI:31291
IUPAC Name bis(oxobismuthanyl) carbonate
SMILES C(=O)(O[Bi]=O)O[Bi]=O

Specifications

Physical Form Powder
Quantity 500 g
Merck Index 14,1282
Solubility Information Practically insoluble in water,alcohol; Soluble in nitric acid,hydrochloric acid,concentrated acetic acid;Soluble in mineral acid and glacial acetic acid; Insoluble in water.
Formula Weight 509.97
Percent Purity ≤98.5%
Density 6.86 g/mL
Chemical Name or Material Bismuth carbonate oxide
EINECSNumber 227-567-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.