Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Chloride, 97%, Spectrum™ Chemical
SDP

Catalog No. 18609088 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-088 12 kg Amber Glass Bottle
18-609-089 25 g Amber Glass Bottle
18-609-087 100 g Amber Glass Bottle
18-609-090 500 g Amber Glass Bottle
18-609-091 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-609-088 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B111012KGBL
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 7787-60-2

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 12 kg
Molecular Weight (g/mol) 315.34
Percent Purity 97%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Bismuth Chloride

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.