Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth Citrate, Powder, USP, 49-54%, Spectrum™ Chemical
SDP

Catalog No. 18600998 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-998 125 g Amber Glass Bottle
18-600-997 25 g Amber Glass Bottle
18-600-999 500 g Amber Glass Bottle
18-601-001 12 kg Amber Glass Bottle
18-601-000 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-998 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1973125GM
Add to Cart
Add to Cart

B1973, 813-93-4, C6H5BiO7

Spectrum™ Chemical Bismuth Citrate, Powder, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 813-93-4
Molecular Formula C6H5BiO7
Molecular Weight (g/mol) 398.08
InChI Key ANERHPOLUMFRDC-UHFFFAOYSA-K
IUPAC Name bismuth(3+) 2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Bi+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 125 g
Percent Purity 49% to 54%
Grade USP
Packaging Amber Glass Bottle
Chemical Name or Material Bismuth Citrate, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.