Learn More
Bismuth hydroxide nitrate oxide, Bi{2}O{3} 79% min
Description
It is used pharmaceutically in wound dressing, ointment component and as an antiacid. In cosmetics it is used as a cosmetic white pigment. Used in Ceramic glazes and enamel fluxes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 260°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
UN Number | UN1477 |
Merck Index | 14,1284 |
Solubility Information | Practically insoluble in water and alcohol; Soluble in dilute HNO3 and HCl |
Formula Weight | 1461.98 |
Concentration or Composition (by Analyte or Components) | Bi2O3 79% min. |
Odor | Odorless |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | H272-H315-H319-H335 |
missing translation for 'healthHazard2' | GHS H Statement H272-H315-H319-H335 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
missing translation for 'healthHazard3' | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S. |
EINECSNumber | 215-136-8 |
RTECSNumber | EB2977000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.