Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bismuth ICP Standard, 10,000 ppm Bi in 5% HNO3, Ricca Chemical

Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
Specifications
Specifications
Packaging | Natural Poly Bottle |
Concentration or Composition Notes | 10,000ppm Bi |
CAS Min % | 86.0, 11.57, 0.86 |
CAS Max % | 89.0, 12.05, 0.89 |
Equivalence | 1 mL = 10 mg Bi |
Physical Form | Liquid |
Color | Colorless |
pH | < 2 |
Type | Standard |
Quantity | 50 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction