Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) bromide, anhydrous, 99.999% (metals basis)

Catalog No. AA4717503
Click to view available options
Quantity:
1 g
5 g

CAS: 7787-58-8 | BiBr3 | 448.692 g/mol

It catalyzes the formation of cyclic carbonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-58-8
Molecular Formula BiBr3
Molecular Weight (g/mol) 448.692
MDL Number MFCD00014195
InChI Key TXKAQZRUJUNDHI-UHFFFAOYSA-K
Synonym bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo
PubChem CID 82232
IUPAC Name tribromobismuthane
SMILES Br[Bi](Br)Br

Specifications

Boiling Point 453°C
Melting Point 218°C
Color Yellow
Physical Form Powder
Quantity 1 g
UN Number UN3260
Solubility Information Decomposes in water.
Formula Weight 448.71
Percent Purity 99.999%
Odor Acidic
Sensitivity Hygroscopic
Density 5.72 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth(III) bromide, Anhydrous
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-121-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.