Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) chloride, 97+%, anhydrous

Catalog No. p-3734660
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 7787-60-2 | BiCl3 | 315.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-60-2
Molecular Formula BiCl3
Molecular Weight (g/mol) 315.33
MDL Number MFCD00003461
InChI Key JHXKRIRFYBPWGE-UHFFFAOYSA-K
Synonym bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride
PubChem CID 24591
IUPAC Name trichlorobismuthane
SMILES Cl[Bi](Cl)Cl

Specifications

Boiling Point 447.0°C
Melting Point 230.0°C to 232.0°C
Color Beige to White or Gray
Physical Form Crystalline Powder and/or Chunks
Quantity 25 g
Assay Percent Range 98% min. (Argentometry)
Linear Formula BiCl3
Fieser 07,24
Merck Index 15, 1264
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in acids, soluble in abs.alcohol, acetone and ethyl acetate
Formula Weight 315.33
Percent Purity ≥97%
Packaging Glass bottle
Specific Gravity 4.75
Density 4.7500g/mL
Chemical Name or Material Bismuth(III) chloride, Anhydrous
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 232-123-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.