Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water

Catalog No. AAA1051022
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 7787-60-2 | BiCl3 | 315.33 g/mol

Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.

Solubility
Soluble in hydrochloric acid, nitric acid, diethyl ether, ethyl acetate and acetone. Insoluble in alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-60-2
Molecular Formula BiCl3
Molecular Weight (g/mol) 315.33
MDL Number MFCD00003461
InChI Key JHXKRIRFYBPWGE-UHFFFAOYSA-K
Synonym bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride
PubChem CID 24591
IUPAC Name trichlorobismuthane
SMILES Cl[Bi](Cl)Cl

Specifications

Boiling Point ∼430°C (sublimation)
Melting Point 230°C to 232°C
Quantity 100 g
UN Number UN3260
Merck Index 14,1262
Solubility Information Soluble in hydrochloric acid,nitric acid,diethyl ether,ethyl acetate and acetone. Insoluble in alcohol.
Formula Weight 315.34
Percent Purity 98%
Concentration or Composition (by Analyte or Components) May contain up to 3% water
Odor Irritating
Sensitivity Moisture sensitive
Density 4.75
Assay (dry wt.)
Chemical Name or Material Bismuth (III) chloride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 232-123-2
RTECSNumber EB2690000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.