Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) chloride oxide, 98%

Catalog No. AA4039630
Click to view available options
Quantity:
50 g
250 g
1 kg

Used as pigment, in manufacture of artificial pearls, dry-cell cathodes | CAS: 7787-59-9 | BiClHO | 261.437 g/mol

Bismuth(III) chloride oxide is used in pharmaceutical industry, in the production of cosmetics, in chemical and metallurgical industries. It is used as pigment, in manufacture of artificial pearls and as a dry-cell cathodes, in photocatalysts for degradation of organic pollutant, in white-light LEDs, in the manufacture of plastics, in the synthesis of methanol, in semiconductor devices such as CCDs or photodiodes and also used in the production of permanent magnets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-59-9
Molecular Formula BiClHO
Molecular Weight (g/mol) 261.437
MDL Number MFCD00010895
InChI Key BWOROQSFKKODDR-UHFFFAOYSA-N
Synonym bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid
PubChem CID 6328152
IUPAC Name oxobismuth;hydrochloride
SMILES O=[Bi].Cl

Specifications

Melting Point (decomposition)
Color Brown
Physical Form Powder
Quantity 250 g
Linear Formula BiOCl
Merck Index 14,1263
Solubility Information Insoluble in water,alcohol; Soluble in acids.
Formula Weight 260.43
Percent Purity 98%
Odor Odorless
Refractive Index 2.15
Density 7.72 g/mL
Chemical Name or Material Bismuth(III) chloride oxide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-122-7
RTECSNumber EB2700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.