Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) fluoride, 98%, Thermo Scientific™

Catalog No. AC311010250
Click to view available options
:
25g; Plastic bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7787-61-3
Quantity 25g
Assay Percent Range 98%
Molecular Formula BiF3
MDL Number MFCD00010893
Merck Index 15, 1265
Synonym bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
InChI Key BRCWHGIUHLWZBK-UHFFFAOYSA-K
SMILES F[Bi](F)F
IUPAC Name trifluorobismuthane
Molecular Weight (g/mol) 265.976
PubChem CID 82233
Formula Weight 265.98
Percent Purity ≥97.5%
Identification Confirmed
Packaging Plastic bottle
Chemical Name or Material Bismuth(III) fluoride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.