Learn More
Bismuth(III) fluoride, anhydrous, 99%
Description
It is used in the manufacturing of bismuth pentafluoride. Bismuth(III) fluoride is an attractive reagent and catalyst in organic synthesis owing to its low cost, and ease of handling. It is also used in polarography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 727°C |
Physical Form | Powder |
Quantity | 500 g |
UN Number | UN3260 |
Merck Index | 14,1265 |
Solubility Information | Insoluble in water; Soluble in HF. |
Formula Weight | 265.98 |
Percent Purity | 99% |
Odor | Odorless |
Refractive Index | 1.74 |
Show More |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H314-H319 Causes severe skin burns and eye damage. Causes serious eye irritation. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 232-124-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.