Learn More
Bismuth(III) iodide, Puratronic™, 99.999% (metals basis)
Description
Polycrystalline films of bismuth triiodide are used as beta direct and digital detectors for medical imaging applications. Bismuth(III) iodide is used as an ice nucleant. It is also used in electron microscopy to reveal synapses. Addition of bismuth triiodide has been shown to improve the glass forming ability and thermal stability of Ge-Te-BiI(3) chalcogenide glasses. Solution-processable BiI3 nanosheets have been shown to exhibit the potential to be used as HTLs (Hole Transport Layer) in conventional OPV (organic photovoltaic) devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | ∼500°C |
Melting Point | 408°C |
Color | Gray to Black |
Physical Form | Lump/Powder |
Quantity | 50 g |
UN Number | UN3260 |
Merck Index | 14,1269 |
Solubility Information | Insoluble in water. Highly soluble in ethanol,benzene,toluene and xylene,hydrochloric acid and in hydriodic acid. |
IUPAC Name | bismuth(3+) triiodide |
Formula Weight | 589.69 |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
EINECSNumber | 232-127-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.