Learn More
Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Description
It is used in electronic ceramics and fluorescent paints. It is employed as an oxidant for selective oxidation of sulfides to sulfoxides, as a nitrating reagent for synthesizing nitro aromatic compounds, and as a precursor for Dragendorff reagent for use in TLC stain and for testing the presence of alkaloids. On silica gel support, it has been reported to be an efficient catalyst for the Biginelli reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 30°C (decomposition) |
Color | Colorless to White |
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN1477 |
Merck Index | 14,1271 |
Solubility Information | Slightly soluble in water; soluble in acetone,acetic acid,and glycerol. Insoluble in ethyl acetate and ethanol. |
Formula Weight | 394.99 (Anhydrous) |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Show More |
Safety and Handling
Hazard Category | H272-H315-H319-H335 |
Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S. |
EINECSNumber | 233-791-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.