Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) nitrate pentahydrate, 99.999%, (trace metal basis)

Catalog No. p-3734662
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Catalog No. AC315150050

Supplier: Thermo Scientific Chemicals 315150050

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 10035-06-0 | BiH10N3O14 | 485.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-06-0
Molecular Formula BiH10N3O14
Molecular Weight (g/mol) 485.07
MDL Number MFCD00149157
InChI Key FBXVOTBTGXARNA-UHFFFAOYSA-N
Synonym bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate
PubChem CID 160911
IUPAC Name bismuth;trinitrate;pentahydrate
SMILES O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 80.0°C
Melting Point 30.0°C
Color White
Physical Form Crystals
Quantity 5 g
Assay Percent Range 99.999%
Linear Formula Bi(NO3)3·5H2O
Merck Index 15, 1271
Solubility Information Solubility in water: may decompose
Formula Weight 485.07
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Bismuth(III) nitrate pentahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.